Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TATTTTAAAAATCCTGAATGGCAAG[G/T]TTTTTGGAATGGTTTAATAACATAG
Species: |
Human | |||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||
Phenotype: |
MIM: 612907 | |||||||||||||||||||||||
Literature Links: |
TRNT1 PubMed Links | |||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR - Not Available | |||||
AMR - Not Available |
TRNT1 - tRNA nucleotidyl transferase 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001302946.1 | Intron | NP_001289875.1 | ||||
NM_182916.2 | Intron | NP_886552.2 | ||||
XM_011533776.2 | Intron | XP_011532078.1 | ||||
XM_011533777.2 | Intron | XP_011532079.1 | ||||
XM_011533778.2 | Intron | XP_011532080.1 |