Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GTCATTTGCTGTGTCTTCTTCCCTC[A/G]TGGTAGAGAGGGTTTGGTCCTACGT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 610892 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
LOC550113 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
LOC550113 - uncharacterized LOC550113 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
SYT14P1 - synaptotagmin 14 pseudogene 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
TMPRSS11F - transmembrane protease, serine 11F | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_207407.2 | 1550 | UTR 3 | NP_997290.2 | |||
XM_011531935.1 | 1550 | Intron | XP_011530237.1 |