Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TCAAGCTGGTGGTCCACGACTGTCC[C/T]GTCTGGGTAAGGAAGAGCAGCTGCT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 607353 MIM: 608039 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
RHOBTB3 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
RHOBTB3 - Rho related BTB domain containing 3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_014899.3 | 759 | Silent Mutation | CCC,CCT | P,P 74 | NP_055714.3 | |
XM_011543279.2 | 759 | Silent Mutation | CCC,CCT | P,P 74 | XP_011541581.1 | |
XM_017009236.1 | 759 | UTR 5 | XP_016864725.1 | |||
XM_017009237.1 | 759 | UTR 5 | XP_016864726.1 |
SPATA9 - spermatogenesis associated 9 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_031952.3 | 759 | Intron | NP_114158.2 | |||
XM_011543664.1 | 759 | Intron | XP_011541966.1 | |||
XM_011543666.1 | 759 | Intron | XP_011541968.1 | |||
XM_011543667.2 | 759 | Intron | XP_011541969.1 | |||
XM_017009948.1 | 759 | Intron | XP_016865437.1 | |||
XM_017009949.1 | 759 | Intron | XP_016865438.1 | |||
XM_017009950.1 | 759 | Intron | XP_016865439.1 | |||
XM_017009951.1 | 759 | Intron | XP_016865440.1 |