Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TGTTGCCCCATGTATGGTTCCATCA[C/T]TGGACCCTTTCCCATCCTTTTGTTA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 604941 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
PPP2R2A PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
PPP2R2A - protein phosphatase 2 regulatory subunit Balpha | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001177591.1 | Intron | NP_001171062.1 | ||||
NM_002717.3 | Intron | NP_002708.1 | ||||
XM_005273559.1 | Intron | XP_005273616.1 | ||||
XM_005273560.1 | Intron | XP_005273617.1 | ||||
XM_011544565.1 | Intron | XP_011542867.1 | ||||
XM_011544566.1 | Intron | XP_011542868.1 | ||||
XM_011544567.2 | Intron | XP_011542869.1 | ||||
XM_011544568.1 | Intron | XP_011542870.1 | ||||
XM_011544569.1 | Intron | XP_011542871.1 |