Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AGACCAGACCCAGAAATTAAAAGTC[A/G]AGAGTGTAACTGTAGTTGAACTAAT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 614285 MIM: 613324 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
C1QTNF9 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
C1QTNF9 - C1q and tumor necrosis factor related protein 9 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001303137.1 | Intron | NP_001290066.1 | ||||
NM_001303138.1 | Intron | NP_001290067.1 | ||||
NM_178540.4 | Intron | NP_848635.2 |
SPATA13 - spermatogenesis associated 13 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |