Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TGTGAGTAGAGCAATTCTAAAAATC[C/T]AGCATGATTTAAGTAGAGTAGGACT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||
Literature Links: |
USP12 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
USP12 - ubiquitin specific peptidase 12 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_182488.3 | Intron | NP_872294.2 | ||||
XM_005266282.2 | Intron | XP_005266339.1 | ||||
XM_011534956.2 | Intron | XP_011533258.1 | ||||
XM_017020416.1 | Intron | XP_016875905.1 |