Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CCTATTGAATCACTAGAAATAGAGA[A/G]GCTTTTAAAAAAATTAACTTGTTTT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 605397 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
CD226 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
CD226 - CD226 molecule | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001303618.1 | 4374 | Intron | NP_001290547.1 | |||
NM_001303619.1 | 4374 | Intron | NP_001290548.1 | |||
NM_006566.3 | 4374 | Intron | NP_006557.2 | |||
XM_005266642.3 | 4374 | UTR 3 | XP_005266699.1 | |||
XM_005266643.3 | 4374 | UTR 3 | XP_005266700.1 | |||
XM_006722374.3 | 4374 | UTR 3 | XP_006722437.1 | |||
XM_017025524.1 | 4374 | UTR 3 | XP_016881013.1 | |||
XM_017025525.1 | 4374 | UTR 3 | XP_016881014.1 | |||
XM_017025526.1 | 4374 | UTR 3 | XP_016881015.1 | |||
XM_017025527.1 | 4374 | UTR 3 | XP_016881016.1 |