Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AGTTGAGAGGTGAAAAATACACCAC[C/T]CTAAGTGGTTAATTTGAGGTAGGAG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 609758 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
NKAIN2 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
NKAIN2 - Na+/K+ transporting ATPase interacting 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001040214.2 | Intron | NP_001035304.1 | ||||
NM_001300737.1 | Intron | NP_001287666.1 | ||||
NM_001300738.1 | Intron | NP_001287667.1 | ||||
NM_001300740.1 | Intron | NP_001287669.1 | ||||
NM_153355.4 | Intron | NP_699186.2 | ||||
XM_011535501.2 | Intron | XP_011533803.1 | ||||
XM_011535502.2 | Intron | XP_011533804.1 | ||||
XM_011535503.2 | Intron | XP_011533805.1 | ||||
XM_017010318.1 | Intron | XP_016865807.1 | ||||
XM_017010319.1 | Intron | XP_016865808.1 |