Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TAAATACAGAGCCATATGTTTTAAT[A/G]AGCCAGCAAGCTAGGGAGGGATTGA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 605437 MIM: 611029 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
PRKCH PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
PRKCH - protein kinase C eta | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_006255.4 | Intron | NP_006246.2 | ||||
XM_011536954.2 | Intron | XP_011535256.1 | ||||
XM_011536955.1 | Intron | XP_011535257.1 | ||||
XM_011536956.1 | Intron | XP_011535258.1 | ||||
XM_011536957.1 | Intron | XP_011535259.1 | ||||
XM_017021458.1 | Intron | XP_016876947.1 | ||||
XM_017021459.1 | Intron | XP_016876948.1 |
TMEM30B - transmembrane protein 30B | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |