Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ATACCTGAGTATGCTTACGGAAAAA[C/T]ACCCCTTGCACCTTCCCAAAAATTT
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 600875 | ||||||||||||||||||||
Literature Links: |
ACYP1 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
ACYP1 - acylphosphatase 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001107.4 | 149 | Silent Mutation | GTA,GTG | V,V 21 | NP_001098.1 | |
NM_001302616.1 | 149 | Silent Mutation | GTA,GTG | V,V 21 | NP_001289545.1 | |
NM_001302617.1 | 149 | Silent Mutation | GTA,GTG | V,V 51 | NP_001289546.1 |
ZC2HC1C - zinc finger C2HC-type containing 1C | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |