Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GAAAGGAGCAACTACCAACATCTGC[C/T]ACAATGTACTGGATCGAAATGTCCA
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 605832 | ||||||||||||||||||||
Literature Links: |
ACSS2 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
ACSS2 - acyl-CoA synthetase short-chain family member 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001076552.2 | 387 | Missense Mutation | CAC,TAC | H,Y 105 | NP_001070020.2 | |
NM_001242393.1 | 387 | Missense Mutation | CAC,TAC | H,Y 10 | NP_001229322.1 | |
NM_018677.3 | 387 | Missense Mutation | CAC,TAC | H,Y 105 | NP_061147.1 | |
XM_005260455.1 | 387 | Missense Mutation | CAC,TAC | H,Y 105 | XP_005260512.1 | |
XM_005260456.1 | 387 | Missense Mutation | CAC,TAC | H,Y 105 | XP_005260513.1 | |
XM_006723826.1 | 387 | Missense Mutation | CAC,TAC | H,Y 10 | XP_006723889.1 | |
XM_011528905.1 | 387 | Missense Mutation | CAC,TAC | H,Y 105 | XP_011527207.1 | |
XM_011528906.1 | 387 | Missense Mutation | CAC,TAC | H,Y 105 | XP_011527208.1 | |
XM_011528907.1 | 387 | Missense Mutation | CAC,TAC | H,Y 105 | XP_011527209.1 | |
XM_011528908.1 | 387 | Missense Mutation | CAC,TAC | H,Y 105 | XP_011527210.1 | |
XM_011528909.2 | 387 | Missense Mutation | CAC,TAC | H,Y 55 | XP_011527211.1 | |
XM_011528911.1 | 387 | Missense Mutation | CAC,TAC | H,Y 55 | XP_011527213.1 | |
XM_011528912.2 | 387 | Missense Mutation | CAC,TAC | H,Y 10 | XP_011527214.1 |