Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ACTCCTGGCAGGCCCTGACTGGCAG[C/T]CAGTGGTGTGACTCTGAGGTGGGCC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 604269 MIM: 606561 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
FAT2 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
FAT2 - FAT atypical cadherin 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001447.2 | 14132 | UTR 3 | NP_001438.1 | |||
XM_006714761.3 | 14132 | UTR 3 | XP_006714824.1 | |||
XM_011537600.2 | 14132 | UTR 3 | XP_011535902.1 | |||
XM_011537603.2 | 14132 | UTR 3 | XP_011535905.1 | |||
XM_017009224.1 | 14132 | UTR 3 | XP_016864713.1 | |||
XM_017009225.1 | 14132 | UTR 3 | XP_016864714.1 | |||
XM_017009226.1 | 14132 | Intron | XP_016864715.1 | |||
XM_017009227.1 | 14132 | Intron | XP_016864716.1 |