Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GCAATGTGTGTTTAAGGCAGAGCCC[A/G]GCAGATCCGTGTCCACCCATGTGCC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 182120 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
SPARC PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
SPARC - secreted protein acidic and cysteine rich | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001309443.1 | 1408 | UTR 3 | NP_001296372.1 | |||
NM_001309444.1 | 1408 | UTR 3 | NP_001296373.1 | |||
NM_003118.3 | 1408 | UTR 3 | NP_003109.1 |