Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AGAATTAAAATAAATTTCTGCTGCT[G/T]CTTCAACATAGTTTAGTAGTATTGT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 616563 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
SLK PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
SLK - STE20 like kinase | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001304743.1 | Intron | NP_001291672.1 | ||||
NM_014720.3 | Intron | NP_055535.2 | ||||
XM_011540401.2 | Intron | XP_011538703.1 |