Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GTGTTACAGCACGCACCTGAGAAGT[C/T]CTAAGAGGCGATTCTATCCAGGTTT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||
Literature Links: |
CFAP58 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
CFAP58 - cilia and flagella associated protein 58 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001008723.1 | Intron | NP_001008723.1 | ||||
XM_011539377.2 | Intron | XP_011537679.1 | ||||
XM_017015787.1 | Intron | XP_016871276.1 | ||||
XM_017015788.1 | Intron | XP_016871277.1 | ||||
XM_017015789.1 | Intron | XP_016871278.1 | ||||
XM_017015790.1 | Intron | XP_016871279.1 |
CFAP58-AS1 - CFAP58 antisense RNA 1 (head to head) | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
ITPRIP - inositol 1,4,5-trisphosphate receptor interacting protein | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |