Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TGTCTCAAATAGAAGATGAAAGTAT[A/G]ATCCTATCCTCCAGATGAAACCATT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
29 submissions
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 606228 MIM: 607355 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
AGO1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
AGO1 - argonaute 1, RISC catalytic component | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
AGO3 - argonaute 3, RISC catalytic component | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_024852.3 | Intron | NP_079128.2 | ||||
NM_177422.2 | Intron | NP_803171.1 | ||||
XM_005270575.3 | Intron | XP_005270632.1 | ||||
XM_005270576.3 | Intron | XP_005270633.1 | ||||
XM_011540879.2 | Intron | XP_011539181.1 | ||||
XM_011540882.2 | Intron | XP_011539184.1 | ||||
XM_017000523.1 | Intron | XP_016856012.1 | ||||
XM_017000524.1 | Intron | XP_016856013.1 | ||||
XM_017000525.1 | Intron | XP_016856014.1 | ||||
XM_017000526.1 | Intron | XP_016856015.1 | ||||
XM_017000527.1 | Intron | XP_016856016.1 | ||||
XM_017000528.1 | Intron | XP_016856017.1 |