Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GACAGTGACAGCCGGACAAGAAGCT[A/G]AAGTGACAGGTGAGAAAACACTGAA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 606436 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
SYT12 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
SYT12 - synaptotagmin 12 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001177880.1 | 913 | Intron | NP_001171351.1 | |||
NM_001318773.1 | 913 | Intron | NP_001305702.1 | |||
NM_001318775.1 | 913 | Intron | NP_001305704.1 | |||
NM_177963.3 | 913 | Intron | NP_808878.1 | |||
XM_006718737.3 | 913 | UTR 5 | XP_006718800.1 | |||
XM_011545346.2 | 913 | UTR 5 | XP_011543648.1 | |||
XM_011545347.1 | 913 | Intron | XP_011543649.1 | |||
XM_017018547.1 | 913 | Intron | XP_016874036.1 | |||
XM_017018548.1 | 913 | Intron | XP_016874037.1 |