Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 607854 | ||||||||||||||||||||
Literature Links: |
BEST1 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
BEST1 - bestrophin 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001139443.1 | 668 | Intron | NP_001132915.1 | |||
NM_001300786.1 | 668 | Intron | NP_001287715.1 | |||
NM_001300787.1 | 668 | Intron | NP_001287716.1 | |||
NM_004183.3 | 668 | Nonsense Mutation | AAG,TAG | K,* 30 | NP_004174.1 |
RAB3IL1 - RAB3A interacting protein like 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |