Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AGAAAAGGAATGGATGGTCGGCAGT[A/G]AAATTTTCTTCGGGCATCAACATGC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 130135 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
SERPINB1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
SERPINB1 - serpin family B member 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_030666.3 | 1436 | Silent Mutation | TTC,TTT | F,F 352 | NP_109591.1 | |
XM_011514333.1 | 1436 | Silent Mutation | TTC,TTT | F,F 352 | XP_011512635.1 | |
XM_011514334.2 | 1436 | Silent Mutation | TTC,TTT | F,F 352 | XP_011512636.1 | |
XM_011514335.2 | 1436 | Silent Mutation | TTC,TTT | F,F 303 | XP_011512637.1 |