Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CCCAAGGAAACAACACCCGTGTGGA[C/T]AAACACATGTGGGCGGCTGGGCCAA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 603453 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
RIPK1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
RIPK1 - receptor interacting serine/threonine kinase 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
XM_005249457.3 | Intron | XP_005249514.2 | ||||
XM_006715237.3 | Intron | XP_006715300.2 | ||||
XM_017011403.1 | Intron | XP_016866892.1 | ||||
XM_017011404.1 | Intron | XP_016866893.1 | ||||
XM_017011405.1 | Intron | XP_016866894.1 |