Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ACCTGGAGCAGCTGCTCTTCAGCCA[C/G]ATGCTGGGTAAGTCCTGCCGCCCCG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
14 submissions
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 611628 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
NAV1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
NAV1 - neuron navigator 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001167738.1 | 776 | Intron | NP_001161210.1 | |||
NM_020443.4 | 776 | Missense Mutation | CAC,CAG | H,Q 250 | NP_065176.3 | |
XM_006711609.2 | 776 | Intron | XP_006711672.2 | |||
XM_006711610.2 | 776 | Intron | XP_006711673.2 | |||
XM_006711611.1 | 776 | Intron | XP_006711674.1 | |||
XM_011510097.1 | 776 | Missense Mutation | CAC,CAG | H,Q 250 | XP_011508399.1 | |
XM_011510098.1 | 776 | Missense Mutation | CAC,CAG | H,Q 250 | XP_011508400.1 | |
XM_011510099.1 | 776 | Missense Mutation | CAC,CAG | H,Q 250 | XP_011508401.1 | |
XM_011510100.1 | 776 | Missense Mutation | CAC,CAG | H,Q 250 | XP_011508402.1 | |
XM_011510101.1 | 776 | Missense Mutation | CAC,CAG | H,Q 250 | XP_011508403.1 | |
XM_011510102.1 | 776 | Missense Mutation | CAC,CAG | H,Q 250 | XP_011508404.1 | |
XM_017002751.1 | 776 | Missense Mutation | CAC,CAG | H,Q 250 | XP_016858240.1 | |
XM_017002752.1 | 776 | Intron | XP_016858241.1 | |||
XM_017002753.1 | 776 | Missense Mutation | CAC,CAG | H,Q 250 | XP_016858242.1 | |
XM_017002754.1 | 776 | Missense Mutation | CAC,CAG | H,Q 250 | XP_016858243.1 |