Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GCTGGTGCTTCCTAGGGGGCTCCCA[C/G]AGGCAGGGCTCATCTGGTAGGGTCC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
8 submissions
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 147522 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
ITPKB PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
ITPKB - inositol-trisphosphate 3-kinase B | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_002221.3 | 3494 | UTR 3 | NP_002212.3 | |||
XM_005273120.2 | 3494 | Intron | XP_005273177.1 | |||
XM_017001211.1 | 3494 | Intron | XP_016856700.1 |