Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ATAAGTTAGTGACTGAGAGATTCAA[G/T]TTTTCGAGGGAAGAATAGGAACAGA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 603415 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
LOC101929680 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
LOC101929680 - uncharacterized LOC101929680 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
SCN9A - sodium voltage-gated channel alpha subunit 9 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_002977.3 | Intron | NP_002968.1 | ||||
XM_005246757.2 | Intron | XP_005246814.1 | ||||
XM_011511616.2 | Intron | XP_011509918.1 | ||||
XM_011511617.2 | Intron | XP_011509919.1 | ||||
XM_011511618.2 | Intron | XP_011509920.1 | ||||
XM_011511619.2 | Intron | XP_011509921.1 | ||||
XM_017004668.1 | Intron | XP_016860157.1 | ||||
XM_017004669.1 | Intron | XP_016860158.1 |