Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GAAATCCTCAGCCAGGAAAGCAGTC[C/G]TAGTTAAAACTTCATGGCGCTTTGT
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||
Literature Links: |
RABL3 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
RABL3 - RAB, member of RAS oncogene family like 3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_173825.3 | 502 | Missense Mutation | ACG,AGG | T,R 163 | NP_776186.2 | |
XM_006713595.2 | 502 | Missense Mutation | ACG,AGG | T,R 163 | XP_006713658.1 | |
XM_017006227.1 | 502 | Missense Mutation | ACG,AGG | T,R 163 | XP_016861716.1 | |
XM_017006228.1 | 502 | Missense Mutation | ACG,AGG | T,R 163 | XP_016861717.1 |