Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TTGTTTGGTAGCTTTTTCTGGTGGT[A/G]GTAGTGTTATTCCTGTGATACAGCC
Species: |
Human | |||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||
Phenotype: |
MIM: 604384 | |||||||||||||||||||||||
Literature Links: |
ATP2C1 PubMed Links | |||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR - Not Available | |||||
AMR - Not Available |
ATP2C1 - ATPase secretory pathway Ca2+ transporting 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001001485.2 | Intron | NP_001001485.1 | ||||
NM_001001486.1 | Intron | NP_001001486.1 | ||||
NM_001001487.1 | Intron | NP_001001487.1 | ||||
NM_001199179.1 | Intron | NP_001186108.1 | ||||
NM_001199180.1 | Intron | NP_001186109.1 | ||||
NM_001199181.1 | Intron | NP_001186110.1 | ||||
NM_001199182.1 | Intron | NP_001186111.1 | ||||
NM_001199183.1 | Intron | NP_001186112.1 | ||||
NM_001199184.1 | Intron | NP_001186113.1 | ||||
NM_001199185.1 | Intron | NP_001186114.1 | ||||
NM_014382.3 | Intron | NP_055197.2 | ||||
XM_005247354.2 | Intron | XP_005247411.1 | ||||
XM_005247355.2 | Intron | XP_005247412.1 | ||||
XM_005247356.2 | Intron | XP_005247413.1 | ||||
XM_005247358.2 | Intron | XP_005247415.1 | ||||
XM_011512686.2 | Intron | XP_011510988.1 | ||||
XM_017006164.1 | Intron | XP_016861653.1 |