Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TACTATTTCTCATTCTCTGTCTGAT[G/T]AATGTCACAAATGTGTAACTTTCCA
Species: |
Human | |||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||
Phenotype: |
MIM: 601589 | |||||||||||||||||||||||
Literature Links: |
RASA2 PubMed Links | |||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR - Not Available | |||||
AMR - Not Available |
RASA2 - RAS p21 protein activator 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001303245.1 | Intron | NP_001290174.1 | ||||
NM_001303246.1 | Intron | NP_001290175.1 | ||||
NM_006506.3 | Intron | NP_006497.2 | ||||
XM_011513059.2 | Intron | XP_011511361.1 | ||||
XM_017006968.1 | Intron | XP_016862457.1 | ||||
XM_017006969.1 | Intron | XP_016862458.1 | ||||
XM_017006970.1 | Intron | XP_016862459.1 | ||||
XM_017006971.1 | Intron | XP_016862460.1 | ||||
XM_017006972.1 | Intron | XP_016862461.1 |