Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CTTGGTAAAGGTAAACTGAAAATCT[A/G]TGATGAAGTTAGAAAATATTGAAAA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 610841 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
LOC105374546 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
LOC105374546 - uncharacterized LOC105374546 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
STIM2 - stromal interaction molecule 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001169117.1 | Intron | NP_001162588.1 | ||||
NM_001169118.1 | Intron | NP_001162589.1 | ||||
NM_020860.3 | Intron | NP_065911.3 |
TBC1D19 - TBC1 domain family member 19 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |