Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TCTTGGAAATCAGAATGTATCTTAC[C/T]GTTGGTGGGTCCCATATAATTGACA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 614315 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
ACOT12 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
ACOT12 - acyl-CoA thioesterase 12 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_130767.2 | 2750 | Intron | NP_570123.1 | |||
XM_006714531.2 | 2750 | Intron | XP_006714594.1 | |||
XM_006714532.3 | 2750 | Intron | XP_006714595.1 | |||
XM_017009045.1 | 2750 | UTR 3 | XP_016864534.1 | |||
XM_017009046.1 | 2750 | UTR 3 | XP_016864535.1 | |||
XM_017009047.1 | 2750 | UTR 3 | XP_016864536.1 | |||
XM_017009048.1 | 2750 | UTR 3 | XP_016864537.1 |
CKMT2-AS1 - CKMT2 antisense RNA 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
ZCCHC9 - zinc finger CCHC-type containing 9 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001131035.1 | 2750 | Intron | NP_001124507.1 | |||
NM_001131036.1 | 2750 | Intron | NP_001124508.1 | |||
NM_032280.2 | 2750 | Intron | NP_115656.1 | |||
XM_005248616.3 | 2750 | Intron | XP_005248673.1 |