Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CAATGCATTTATCAATTACCAAAAA[C/T]AGACTAAAGAACAGAAATAAAAGGA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 610847 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
ZNF322 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
ZNF322 - zinc finger protein 322 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001242797.1 | Intron | NP_001229726.1 | ||||
NM_001242798.1 | Intron | NP_001229727.1 | ||||
NM_001242799.1 | Intron | NP_001229728.1 | ||||
NM_024639.4 | Intron | NP_078915.2 |