Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TATTTATGGATCAGAGTGGGCTGGG[A/G]AGAAACACAGCAGCTGATGCCTGTC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 606512 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
PACSIN1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
PACSIN1 - protein kinase C and casein kinase substrate in neurons 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001199583.2 | Intron | NP_001186512.1 | ||||
NM_020804.4 | Intron | NP_065855.1 | ||||
XM_011514541.1 | Intron | XP_011512843.1 |