Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GCCAACAGCCTGACTTCCTCAACCC[A/G]TGGTTCACTCCAGACTCAGTGACCG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 179605 MIM: 609134 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
PRPH2 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
PRPH2 - peripherin 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_000322.4 | 2044 | UTR 3 | NP_000313.2 |
UBR2 - ubiquitin protein ligase E3 component n-recognin 2 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |