Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GCAGAACACTGTTGAAGGCTAGAGA[C/T]TTCAGCTTCATATAGCTACCCCAAA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||
Literature Links: |
GFOD1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
GFOD1 - glucose-fructose oxidoreductase domain containing 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001242628.1 | 4254 | Intron | NP_001229557.1 | |||
NM_001242629.1 | 4254 | Intron | NP_001229558.1 | |||
NM_001242630.1 | 4254 | Intron | NP_001229559.1 | |||
NM_018988.3 | 4254 | Intron | NP_061861.1 | |||
XM_011514699.2 | 4254 | UTR 3 | XP_011513001.1 |