Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ACACCGATTTATTGAGTAGCAGAGC[A/T]GAGGGAAGGCGTAGGATGGCTCCAG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 607929 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
CCM2 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
CCM2 - CCM2 scaffolding protein | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
LOC100128364 - uncharacterized LOC100128364 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
NACAD - NAC alpha domain containing | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001146334.1 | 4639 | UTR 3 | NP_001139806.1 | |||
XM_006715674.3 | 4639 | UTR 3 | XP_006715737.1 |