Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AAAGGAGGAAGCAGTCAGGTATAGG[C/T]CATCTGTGTGTTTGGAGCCTGGGTG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 191730 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
UPP1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
UPP1 - uridine phosphorylase 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001287426.1 | Intron | NP_001274355.1 | ||||
NM_001287428.1 | Intron | NP_001274357.1 | ||||
NM_001287429.1 | Intron | NP_001274358.1 | ||||
NM_001287430.1 | Intron | NP_001274359.1 | ||||
NM_003364.3 | Intron | NP_003355.1 | ||||
XM_005249838.4 | Intron | XP_005249895.1 | ||||
XM_011515512.2 | Intron | XP_011513814.1 | ||||
XM_011515513.2 | Intron | XP_011513815.1 | ||||
XM_011515514.2 | Intron | XP_011513816.1 | ||||
XM_011515515.2 | Intron | XP_011513817.1 |