Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ACCGCGACCCCAGAAGATGCAAGGA[C/G]AGAAGCCCATGAGCTTCTTGAAGGT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 610184 MIM: 603066 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
MOGAT3 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
MOGAT3 - monoacylglycerol O-acyltransferase 3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001287147.1 | 957 | Intron | NP_001274076.1 | |||
NM_178176.3 | 957 | Missense Mutation | TCT,TGT | S,C 263 | NP_835470.1 | |
XM_005250309.3 | 957 | Missense Mutation | TCT,TGT | S,C 263 | XP_005250366.1 | |
XM_011516126.2 | 957 | Missense Mutation | TCT,TGT | S,C 227 | XP_011514428.1 | |
XM_011516127.2 | 957 | Missense Mutation | TCT,TGT | S,C 145 | XP_011514429.1 |
PLOD3 - procollagen-lysine,2-oxoglutarate 5-dioxygenase 3 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |