Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TGGCGTCCTCTGGCCACACGCTCGG[A/G]GGCCCAAGAGGGGTCTCCAGGAAGA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||
Literature Links: |
LINC00051 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
LINC00051 - long intergenic non-protein coding RNA 51 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
TSNARE1 - t-SNARE domain containing 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001291931.1 | Intron | NP_001278860.1 | ||||
NM_145003.4 | Intron | NP_659440.2 | ||||
XM_011516912.1 | Intron | XP_011515214.1 | ||||
XM_011516913.2 | Intron | XP_011515215.1 | ||||
XM_011516914.1 | Intron | XP_011515216.1 | ||||
XM_011516915.2 | Intron | XP_011515217.1 | ||||
XM_011516916.1 | Intron | XP_011515218.1 | ||||
XM_011516917.2 | Intron | XP_011515219.1 | ||||
XM_011516918.1 | Intron | XP_011515220.1 | ||||
XM_011516919.1 | Intron | XP_011515221.1 | ||||
XM_011516920.1 | Intron | XP_011515222.1 | ||||
XM_011516921.1 | Intron | XP_011515223.1 | ||||
XM_011516922.1 | Intron | XP_011515224.1 | ||||
XM_011516923.2 | Intron | XP_011515225.1 | ||||
XM_011516924.1 | Intron | XP_011515226.1 | ||||
XM_017013175.1 | Intron | XP_016868664.1 | ||||
XM_017013176.1 | Intron | XP_016868665.1 | ||||
XM_017013177.1 | Intron | XP_016868666.1 | ||||
XM_017013178.1 | Intron | XP_016868667.1 | ||||
XM_017013179.1 | Intron | XP_016868668.1 | ||||
XM_017013180.1 | Intron | XP_016868669.1 | ||||
XM_017013181.1 | Intron | XP_016868670.1 |