Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GTCACACTCTGCTAGGACAGATGGA[A/G]TAGCCAATGCCAAGAACTTCGCCGG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 605266 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
JPH1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
JPH1 - junctophilin 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001317830.1 | 2107 | UTR 3 | NP_001304759.1 | |||
NM_020647.3 | 2107 | UTR 3 | NP_065698.1 | |||
XM_005251274.3 | 2107 | Intron | XP_005251331.1 | |||
XM_005251275.4 | 2107 | UTR 3 | XP_005251332.1 |