Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CTTGTAGGACTAGATAGAGGCAACA[A/G]TGTCTCGCAAAGGGCAAAATTGTGC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 612488 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
RNF38 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
RNF38 - ring finger protein 38 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_022781.4 | 3621 | UTR 3 | NP_073618.3 | |||
NM_194328.2 | 3621 | UTR 3 | NP_919309.1 | |||
NM_194329.2 | 3621 | UTR 3 | NP_919310.1 | |||
NM_194330.2 | 3621 | UTR 3 | NP_919311.1 | |||
NM_194332.2 | 3621 | UTR 3 | NP_919313.1 | |||
XM_005251366.3 | 3621 | UTR 3 | XP_005251423.1 | |||
XM_005251367.3 | 3621 | UTR 3 | XP_005251424.1 | |||
XM_005251368.3 | 3621 | UTR 3 | XP_005251425.1 | |||
XM_006716721.3 | 3621 | UTR 3 | XP_006716784.1 | |||
XM_011517712.2 | 3621 | UTR 3 | XP_011516014.1 | |||
XM_011517713.2 | 3621 | UTR 3 | XP_011516015.1 | |||
XM_017014294.1 | 3621 | UTR 3 | XP_016869783.1 | |||
XM_017014295.1 | 3621 | UTR 3 | XP_016869784.1 | |||
XM_017014296.1 | 3621 | UTR 3 | XP_016869785.1 | |||
XM_017014297.1 | 3621 | UTR 3 | XP_016869786.1 |