Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TGTTCTGGTGAGCATGTAGCCCTCA[A/C]CAGAAGCAGGCCAGATGCAGTACTA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||
Literature Links: |
SUSD1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
SUSD1 - sushi domain containing 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001282640.1 | Intron | NP_001269569.1 | ||||
NM_001282643.1 | Intron | NP_001269572.1 | ||||
NM_022486.4 | Intron | NP_071931.2 | ||||
XM_011518915.1 | Intron | XP_011517217.1 | ||||
XM_011518916.2 | Intron | XP_011517218.1 | ||||
XM_011518917.1 | Intron | XP_011517219.1 | ||||
XM_011518918.1 | Intron | XP_011517220.1 | ||||
XM_011518919.1 | Intron | XP_011517221.1 | ||||
XM_017015037.1 | Intron | XP_016870526.1 | ||||
XM_017015038.1 | Intron | XP_016870527.1 | ||||
XM_017015039.1 | Intron | XP_016870528.1 | ||||
XM_017015040.1 | Intron | XP_016870529.1 |