Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TCTTCCATATCCTCTACTTCATCCT[C/T]GTGTATCTTCAAGGCTCTCACCATT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 604503 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
JMJD1C PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
JMJD1C - jumonji domain containing 1C | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001282948.1 | 7819 | Missense Mutation | AAG,GAG | K,E 2349 | NP_001269877.1 | |
NM_001318153.1 | 7819 | Missense Mutation | AAG,GAG | K,E 2243 | NP_001305082.1 | |
NM_001318154.1 | 7819 | Missense Mutation | AAG,GAG | K,E 2349 | NP_001305083.1 | |
NM_001322252.1 | 7819 | Missense Mutation | AAG,GAG | K,E 2493 | NP_001309181.1 | |
NM_001322254.1 | 7819 | Missense Mutation | AAG,GAG | K,E 2312 | NP_001309183.1 | |
NM_001322258.1 | 7819 | Missense Mutation | AAG,GAG | K,E 2312 | NP_001309187.1 | |
NM_032776.2 | 7819 | Missense Mutation | AAG,GAG | K,E 2531 | NP_116165.1 | |
XM_011539503.1 | 7819 | Missense Mutation | AAG,GAG | K,E 2308 | XP_011537805.1 | |
XM_011539508.2 | 7819 | Intron | XP_011537810.1 | |||
XM_017015897.1 | 7819 | Missense Mutation | AAG,GAG | K,E 2349 | XP_016871386.1 | |
XM_017015898.1 | 7819 | Missense Mutation | AAG,GAG | K,E 2349 | XP_016871387.1 | |
XM_017015899.1 | 7819 | Missense Mutation | AAG,GAG | K,E 2243 | XP_016871388.1 | |
XM_017015900.1 | 7819 | Missense Mutation | AAG,GAG | K,E 2243 | XP_016871389.1 | |
XM_017015901.1 | 7819 | Missense Mutation | AAG,GAG | K,E 2243 | XP_016871390.1 | |
XM_017015902.1 | 7819 | Missense Mutation | AAG,GAG | K,E 2243 | XP_016871391.1 | |
XM_017015903.1 | 7819 | Missense Mutation | AAG,GAG | K,E 2243 | XP_016871392.1 |