Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GGACCTTATGTAAAACAAAGGTTGG[A/G]CAAAGGTAGTCAGAAAAAAATAGTA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 608517 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
MYPN PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
MYPN - myopalladin | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001256267.1 | Intron | NP_001243196.1 | ||||
NM_001256268.1 | Intron | NP_001243197.1 | ||||
NM_032578.3 | Intron | NP_115967.2 | ||||
XM_017016833.1 | Intron | XP_016872322.1 | ||||
XM_017016834.1 | Intron | XP_016872323.1 |