Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GTTTCCCTGTATCACTGTAGGTCAT[A/G]ACTCCAAGCTAAAACCAAAAGGAAG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 607472 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
YME1L1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
YME1L1 - YME1 like 1 ATPase | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001253866.1 | 2593 | Silent Mutation | GTC,GTT | V,V 610 | NP_001240795.1 | |
NM_014263.3 | 2593 | Silent Mutation | GTC,GTT | V,V 643 | NP_055078.1 | |
NM_139312.2 | 2593 | Silent Mutation | GTC,GTT | V,V 700 | NP_647473.1 | |
XM_011519300.2 | 2593 | Silent Mutation | GTC,GTT | V,V 667 | XP_011517602.1 |