Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TTTTGGTTTTCCCCGACAGCCGCCC[C/T]GCTAAGTAAGACCCCCAGAGCATCA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 600228 MIM: 191190 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
SCNN1A PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
SCNN1A - sodium channel epithelial 1 alpha subunit | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001038.5 | Intron | NP_001029.1 | ||||
NM_001159575.1 | Intron | NP_001153047.1 | ||||
NM_001159576.1 | Intron | NP_001153048.1 |
TNFRSF1A - TNF receptor superfamily member 1A | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |