Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TTTTTGAATATATTTACAATATAAA[C/T]ACTAATTTGTTTCCAAAGTACATAT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||
Literature Links: |
FAM214A PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
FAM214A - family with sequence similarity 214 member A | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001286495.1 | 4197 | UTR 3 | NP_001273424.1 | |||
NM_019600.3 | 4197 | UTR 3 | NP_062546.2 | |||
XM_005254547.2 | 4197 | UTR 3 | XP_005254604.1 | |||
XM_005254548.2 | 4197 | UTR 3 | XP_005254605.1 | |||
XM_011521789.2 | 4197 | UTR 3 | XP_011520091.1 | |||
XM_011521790.1 | 4197 | UTR 3 | XP_011520092.1 |