Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ATTTTCTAATTCTTAAATCCCAGCA[C/T]AAGCCAAAAAGAGAAAGAGAAAAAA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 609984 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
LCTL PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
LCTL - lactase like | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001278562.1 | 2156 | UTR 3 | NP_001265491.1 | |||
NM_207338.3 | 2156 | UTR 3 | NP_997221.2 | |||
XM_017021999.1 | 2156 | Intron | XP_016877488.1 |
ZWILCH - zwilch kinetochore protein | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001287821.1 | 2156 | UTR 3 | NP_001274750.1 | |||
NM_001287822.1 | 2156 | UTR 3 | NP_001274751.1 | |||
NM_001287823.1 | 2156 | UTR 3 | NP_001274752.1 | |||
NM_017975.4 | 2156 | UTR 3 | NP_060445.3 |