Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ATACTATGTGTCTGACATTATGCTA[A/G]GCACATTTGCATACTTTATATCATT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 606881 MIM: 600477 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
FHOD1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
FHOD1 - formin homology 2 domain containing 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
SLC9A5 - solute carrier family 9 member A5 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001323971.1 | Intron | NP_001310900.1 | ||||
NM_001323972.1 | Intron | NP_001310901.1 | ||||
NM_001323973.1 | Intron | NP_001310902.1 | ||||
NM_001323974.1 | Intron | NP_001310903.1 | ||||
NM_001323975.1 | Intron | NP_001310904.1 | ||||
NM_004594.2 | Intron | NP_004585.1 | ||||
XM_017023594.1 | Intron | XP_016879083.1 |