Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CATAAGCCAGGTGGGGCTTCTCCAC[C/T]GGCCTCCAACGGGTGGGCAGGCCCT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 610791 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
SLC43A2 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
SLC43A2 - solute carrier family 43 member 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001284498.1 | 2427 | UTR 3 | NP_001271427.1 | |||
NM_001284499.1 | 2427 | UTR 3 | NP_001271428.1 | |||
NM_001321364.1 | 2427 | UTR 3 | NP_001308293.1 | |||
NM_001321365.1 | 2427 | UTR 3 | NP_001308294.1 | |||
NM_152346.2 | 2427 | UTR 3 | NP_689559.1 | |||
XM_017024177.1 | 2427 | UTR 3 | XP_016879666.1 | |||
XM_017024178.1 | 2427 | UTR 3 | XP_016879667.1 | |||
XM_017024179.1 | 2427 | UTR 3 | XP_016879668.1 |