Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GGGGGCGCTCCAGGGGCCTGCTGCA[C/T]CCTCTGGCCCTCTGCCTCCCCACAG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 605448 MIM: 610820 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
RUNDC3A PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
RUNDC3A - RUN domain containing 3A | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001144825.1 | Intron | NP_001138297.1 | ||||
NM_001144826.1 | Intron | NP_001138298.1 | ||||
NM_006695.4 | Intron | NP_006686.1 | ||||
XM_005256964.3 | Intron | XP_005257021.1 | ||||
XM_017024035.1 | Intron | XP_016879524.1 | ||||
XM_017024036.1 | Intron | XP_016879525.1 | ||||
XM_017024037.1 | Intron | XP_016879526.1 | ||||
XM_017024038.1 | Intron | XP_016879527.1 | ||||
XM_017024039.1 | Intron | XP_016879528.1 | ||||
XM_017024040.1 | Intron | XP_016879529.1 |
RUNDC3A-AS1 - RUNDC3A antisense RNA 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
SLC25A39 - solute carrier family 25 member 39 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |