Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ACTGGCACAAAGTACAGTTGTGCCA[A/G]TAAAGGTTGTAGAACCAATGTGGTG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 602864 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
WNT9B PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
WNT9B - Wnt family member 9B | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001320458.1 | Intron | NP_001307387.1 | ||||
NM_003396.2 | Intron | NP_003387.1 | ||||
XM_011525178.2 | Intron | XP_011523480.1 |