Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TGGCGGGAAGGAGGGAGGCGCGGCT[A/G]GGGAGAGCGCTCGGGAGCTGCCGGG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 616990 MIM: 188350 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
CLUL1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
CLUL1 - clusterin like 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
TYMS - thymidylate synthetase | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001071.2 | 340 | Intron | NP_001062.1 |
TYMSOS - TYMS opposite strand | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001012716.2 | 340 | Nonsense Mutation | CAG,TAG | Q,* 83 | NP_001012734.2 |